Orders LinkContactUsLinkComponent
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Vacuum Controller V-855 for R-210/R-215
Catalogus # 531-0234
Leverancier:   BUCHI
Vacuum Controller, V-850/V-855
Catalogus # 531-0234
Leverancier:  BUCHI
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Vacuum Controller V-855 for R-210/R-215
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option of reducing their water consumption by adding a cooling water valve.
V-855 vacuum controller:
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
Delivery information: Delivered with an air vent valve and pressure sensors.
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
  • Omschrijving:
    Vacuum Controller V-855 for R-210/R-215
  • Seq:
    05
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Communicatiekabel voor R-210/R-215
    Communicatiekabel voor R-210/R-215
    181-0138
    Log in voor beschikbaarheid
    1 * 1 ST - 34,90 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Woulfse fles voor V-850/V-855
    Woulfse fles voor V-850/V-855
    181-0141
    Log in voor beschikbaarheid
    1 * 1 ST - 237,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Automatic distillation probe for V-855
    Automatic distillation probe for V-855
    531-0181
    Log in voor beschikbaarheid
    1 * 1 ST - 250,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Assembly set for R-210/R-215
    Assembly set for R-210/R-215
    531-0236
    Log in voor beschikbaarheid
    1 * 1 ST - 76,80 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Afstandsbediening voor V-850/V-855
    Afstandsbediening voor V-850/V-855
    531-0237
    Log in voor beschikbaarheid
    1 * 1 ST - 669,00 €