pH and ORP electrodes
Glazen pH-elektrode, Ag/AgCl-referentiesysteem, bolvormig membraan, S-glas, nulpunt pH = 7,0, S 1181 HD, 2 - 14, +10...+135 °C, Glas, 12×120 mm, Schroefstekker
Catalogus # SCOI285111483
Leverancier: SI Analytics
Ga naar:
Product Details & Documents
pH electrodes with glass body.
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Specificaties
- Aansluiting:Schroefstekker
- Ø×L:12×120 mm
- Type:S 1181 HD
- Omschrijving:Glazen pH-elektrode, Ag/AgCl-referentiesysteem, bolvormig membraan, S-glas, nulpunt pH = 7,0
- Materiaal:Glas
- Seq:09
- Temperatuur:+10...+135 °C
- pH:2 - 14
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



