Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Calciumchloride 90-98%, TECHNICAL

Calciumchloride 90-98%, TECHNICAL

Plastic bucket, 25 kg
Catalogus # 22313.460
Irriterend
Leverancier:   VWR Chemicals
Calciumchloride 90-98%, TECHNICAL
Catalogus # 22313.460
Leverancier:  VWR Chemicals
CAS-nummer:  10043-52-4
Calciumchloride 90-98%, TECHNICAL
Calciumchloride 90-98%, TECHNICAL

Plastic bucket, 25 kg

CAS-nummer:  10043-52-4
Price
269,00 €
Log in voor beschikbaarheid
1 * 25 kg
Product Details & Documents
Synoniemen:  Chloro calcium, Calcium dichloride, CaCl2
  • CAS-nummer:10043-52-4
  • Formule:CaCl₂
  • MDL-nummer:MFCD00010903
  • Molecular Weight:110.98 g/mol
  • Merck index:14,01659
  • Kokend Pt:1600 °C (1013 hPa)
  • Smelten Pt:772 °C
  • Dichtheid:2,174 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:233-140-8
  • BEREIKEN:01-2119494219-28
Specificaties

Specificatie testresultaten

  • Assay:
    90 - 98 %

Specificaties

  • Verpakking:
    Plastic bucket
  • VPE:
    25 kg

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.