Trays, general purpose
3500 ml, 232×312×55 mm
Catalogus # 216-0809
Leverancier: SciLabware
Ga naar:
Product Details & Documents
PYREX®, borosilicate glass.
- Chemically resistant
- Can be autoclaved and used in microwave ovens
- With handles
Ideal for numerous applications, drying powders and crystals, staining and development, collection of drips and spills from solutions bottles, gel electrophoresis, paper chromatography and dissection work.
Specificaties
- Capaciteit:3500 ml
- Seq:03
- b×d×h:232×312×55 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



