Temperatuur stick, testo 905
Penetration thermometer, testo 905-T1, −50…+350 °C, ±1 °C, 0,1 °C, 200L×3Ø mm, point 4L mm , 70×40×270 mm, 80 g
Catalogus # 620-3313
Leverancier: TESTO
Ga naar:
Product Details & Documents
Mini penetration and surface digital thermometers with rotatable display.
- Automatic off function
- Industrial sensor, K-type thermocouple
- Easy readout of value thanks to the rotatable display
The testo 905 is a fast mini thermometer with a broad measuring range. The 905-T1 has a professional industrial sensor (thermocouple K-type), which has an excellent level of accuracy, especially in the upper measuring range. The sprung, wide thermocouple measuring head of the 905-T2 guarantees a very quick reaction time, and high accuracy level since it always lies flat on the uneven surface of the object being measured.
* Temperature up to 500 °C - short-term.
Levering: Supplied with an attachment clip and AAA batteries for 1000 hours of use.
Diensten: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Specificaties
- Meetbereik:−50…+350 °C
- Resolutie:0,1 °C
- Gewicht:80 g
- Omschrijving:Penetration thermometer, testo 905-T1
- b×d×h:70×40×270 mm
- Juistheid:±1 °C
- Voeler:200L×3Ø mm, point 4L mm
- Cat. No.:620-3313
- Type accu's:3 AAA micro batteries
- Gebruiksduur accu's:1000 h
- Aflezing:LCD