Cavity slides
Cavity slides, helder, 1, 1,5 mm
Catalogus # HECH42410010
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
Cavity microscopy slides.
- Ground edges
- Made from clear glass
Suitable for staining applications in microscopy.
Specificaties
- Aant. wells:1
- Kleur:helder
- Omschrijving:Cavity slides
- Seq:01
- Dikte:1,5 mm