Cold traps, Type KF
Cold trap, KF 29-OK, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
Catalogus # KGWA1735
Leverancier: KGW
Ga naar:
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
- Easy to clean
- No stand material for holding the cold trap
- Protective casing of Dewar flask made of blue coated metal or aluminium
Diensten: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
- Omschrijving:Cold trap, KF 29-OK, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
- Seq:02
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Plastic ring for cold trap, type KF
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Cooling finger S 29-GL