Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Total hardness, model 5-B, 17 - 510 mg/l, 1 KIT
Catalogus # HACH145300
Leverancier:   Hach
Test kits, drop count tests
Catalogus # HACH145300
Leverancier:  Hach
CAS-nummer:  
Test kits, drop count tests
Test kits, drop count tests
Total hardness, model 5-B, 17 - 510 mg/l, 1 KIT
CAS-nummer:  
Price
41,50 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Let op: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    Total hardness, model 5-B
  • Meetbereik:
    17 - 510 mg/l
Specificaties