Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 8% in aqueous solution, extra pure
Plastic bottle, 2,5 L
  KRAF03137.4000
Natriumhydroxide 8% in aqueous solution, extra pure
  KRAF03137.4000
 :  1310-73-2
Natriumhydroxide 8% in aqueous solution, extra pure
Plastic bottle, 2,5 L
 :  1310-73-2
1 * 2,5 L
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :119…135 °C (1013 hPa)
  •  :9…10 °C
  •  :1,53 g/cm³ (20 °C)
  •  :Kamertemperatuur
  •  :215-185-5
  •  :1824
  •  :8,II
  • Verpakking:
    Plastic bottle
  • VPE:
    2,5 L