Natriumhydroxide 8% in aqueous solution, extra pure
Plastic bottle, 2,5 L
KRAF03137.4000
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :119…135 °C (1013 hPa)
- :9…10 °C
- :1,53 g/cm³ (20 °C)
- :Kamertemperatuur
- :215-185-5
- :1824
- :8,II
- Verpakking:Plastic bottle
- VPE:2,5 L