Sample spoons, mini, SP Bel-Art
0,50 ml, PS
Catalogus # BELAF367210050
Leverancier: Bel-Art Products, a Part of SP
Sample spoons, mini, SP Bel-Art
Catalogus # BELAF367210050
Leverancier: Bel-Art Products, a Part of SP
Ga naar:
Product Details & Documents
PS, white.
- Disposable, made from white high-impact PS
- Ideal for collecting specific small volumes of powders and other dry materials
- 0,10 / 0,50 / 1,0 ml models have a rounded bowl for easier emptying of the spoon and a thinner neck that minimises extra material collection
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Capaciteit:0,50 ml
- Materiaal:PS
- Seq:04