Orders LinkContactUsLinkComponent
Objectives for CKX53
LC achromatic objective with 1 mm correction, Ph 40×/0,55, 2,2 mm
Catalogus # OLYMN1477700
Objectives for CKX53
Catalogus # OLYMN1477700
Leverancier:  
Objectives for CKX53
Objectives for CKX53
LC achromatic objective with 1 mm correction, Ph 40×/0,55, 2,2 mm
Discontinued
Product Details & Documents
Specificaties
  • Vergroting:
    Ph 40×/0,55
  • Omschrijving:
    LC achromatic objective with 1 mm correction
  • Werkafstand:
    2,2 mm