Pasteurpipetten
Ungraduated, 10,0 ml, 45 µl, 300 mm, -, Bulk
Catalogus # 612-5473
Leverancier: RATIOLAB
Ga naar:
Product Details & Documents
PE, voor eenmalig gebruik.
- Soft, elastic bulb
- Unbreakable, reduced risk of injury compared to glass
- Inert to biological fluids and most acids
- Graduated pipettes with clearly visible embossed graduations
- Sterile pipettes are gas-sterilised and individually packed
- Suitable for food contact
Specificaties
- Verpakt:Bulk
- Steriel:-
- Capaciteit:10,0 ml
- Omschrijving:Ungraduated
- Lengte:300 mm
- Seq:18
- Druppelvolume:45 µl