Orders LinkContactUsLinkComponent
Face shields
Face shields
Antiwasemlaag, anti-UV (UV 380), Celluloseacetaat, 195×540 mm
Catalogus # UVEX9708.514
Leverancier:   UVEX
Face shields
Catalogus # UVEX9708.514
Leverancier:  UVEX
Face shields
Face shields
Antiwasemlaag, anti-UV (UV 380), Celluloseacetaat, 195×540 mm
Price
47,10 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
  • Twist ratchet enables simple and reliable adjustment to any head size
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement
Certificeringen: EN 166
Documents
Specificaties
  • Type:
    Antiwasemlaag, anti-UV (UV 380)
  • l×b:
    195×540 mm
  • Materiaal:
    Celluloseacetaat
Accessories
Productinformatie
  • Catalogus #
    Materiaal
    Beschikbaarheid
    Price
  • Replacement visor lens
    Replacement visor lens
    111-2543
    Celluloseacetaat
    Log in voor beschikbaarheid
    1 * 1 ST - 23,90 €
Productinformatie
  • Catalogus #
    Materiaal
    Beschikbaarheid
    Price
  • Replacement visor lens, anti-fog
    Replacement visor lens, anti-fog
    111-8229
    Celluloseacetaat
    Log in voor beschikbaarheid
    1 * 1 ST - 33,00 €