Orders LinkContactUsLinkComponent
Hoffmann clips
Hoffmann clips
20 mm
Catalogus # MURRL141/02
Leverancier:   MURRAY S
Hoffmann clips
Catalogus # MURRL141/02
Leverancier:  MURRAY S
Hoffmann clips
20 mm
Price
17,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Bunsen screw clip, nickel plated brass fitted with protection plate and hinged bottom bar.
  • Spanning width = Width of gate
Specificaties
  • Spanwijdte:
    20 mm
  • Seq:
    02