Orders LinkContactUsLinkComponent
Filter candles
Filter candles
0, 13×25 mm
Catalogus # LENZ07206025
Leverancier:   Lenz Laborglas
Filter candles
Catalogus # LENZ07206025
Leverancier:  Lenz Laborglas
Filter candles
Filter candles
0, 13×25 mm
Price
38,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
Specificaties
  • Ø×h:
    13×25 mm
  • Porositeit:
    0
  • Seq:
    06