Filterkroezen
30 ml, 3, 30 mm
Catalogus # LENZ07185330
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Filtration crucibles made of high quality borosilicate glass 3,3.
Specificaties
- Ø plaat:30 mm
- Porositeit:3
- Capaciteit:30 ml
- Seq:10