Filterkroezen
15 ml, 3, 23 mm
Catalogus # LENZ07185323
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Filtration crucibles made of high quality borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specificaties
- Ø plaat:23 mm
- Porositeit:3
- Capaciteit:15 ml
- Seq:09