Filterkroezen
50 ml, 2, 40 mm
Catalogus # LENZ07185240
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Filtration crucibles made of high quality borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
BSE/TSE-certificaten
Specificaties
- Ø plaat:40 mm
- Porositeit:2
- Capaciteit:50 ml
- Seq:07