Filterkroezen
8 ml, 2, 20 mm
Catalogus # LENZ07185220
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Filtration crucibles made of high quality borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
BSE/TSE-certificaten
Specificaties
- Ø plaat:20 mm
- Porositeit:2
- Capaciteit:8 ml
- Seq:04