Potterbuizen
50 ml, 32×195 mm
Catalog # KART6150
Supplier: KARTELL
Jump to:
Product Details & Documents
: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specifications
- Ø inw.×h:32×195 mm
- Capaciteit:50 ml
- Seq:06