Orders LinkContactUsLinkComponent
Twisselmann-extractors
Twisselmann-extractors
Alleen kop, 100 ml, 45/40 NS
Catalog # LENZ05415137
Supplier:   Lenz Laborglas
Twisselmann-extractors
Catalog # LENZ05415137
Supplier:  Lenz Laborglas
Twisselmann-extractors
Twisselmann-extractors
Alleen kop, 100 ml, 45/40 NS
Price
200,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Documents
Specifications
  • Extractorinhoud:
    100 ml
  • Huls:
    45/40 NS
  • Type:
    Alleen kop