Twisselmann-extractors
Alleen kop, 100 ml, 45/40 NS
Catalog # LENZ05415137
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Documents
BSE TSE Glass Columns
Certificats BSE/TSE
Specifications
- Extractorinhoud:100 ml
- Huls:45/40 NS
- Type:Alleen kop