Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Twisselmann-extractors

Compleet, 150 ml, 250 ml, 45/40 NS
Catalogus # LENZ05380043
Leverancier:   Lenz Laborglas
Twisselmann-extractors
Catalogus # LENZ05380043
Leverancier:  Lenz Laborglas
Twisselmann-extractors
Twisselmann-extractors

Compleet, 150 ml, 250 ml, 45/40 NS

Price
765,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance
Specificaties
  • Extractorinhoud:
    150 ml
  • Huls:
    45/40 NS
  • Type:
    Compleet
  • Kolfinhoud:
    250 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.