Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Twisselmann-extractors

Twisselmann-extractors

Compleet, 100 ml, 250 ml, 45/40 NS
Catalogus # LENZ05380037
Leverancier:   Lenz Laborglas
Twisselmann-extractors
Catalogus # LENZ05380037
Leverancier:  Lenz Laborglas
Twisselmann-extractors
Twisselmann-extractors

Compleet, 100 ml, 250 ml, 45/40 NS

Price
753,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance

Documents

Specificaties
  • Extractorinhoud:
    100 ml
  • Huls:
    45/40 NS
  • Type:
    Compleet
  • Kolfinhoud:
    250 ml