Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Twisselmann-extractors

Twisselmann-extractors

Compleet, 70 ml, 100 ml, 34/35 NS
Catalogus # LENZ05380031
Leverancier:   Lenz Laborglas
Twisselmann-extractors
Catalogus # LENZ05380031
Leverancier:  Lenz Laborglas
Twisselmann-extractors
Twisselmann-extractors

Compleet, 70 ml, 100 ml, 34/35 NS

Price
633,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance

Documents

Specificaties
  • Extractorinhoud:
    70 ml
  • Huls:
    34/35 NS
  • Type:
    Compleet
  • Kolfinhoud:
    100 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.