Orders LinkContactUsLinkComponent
Condensators, Davies
Condensators, Davies
250 mm, 24/29 NS, 24/29 NS
Catalogus # LENZ05215313
Leverancier:   Lenz Laborglas
Condensators, Davies
Catalogus # LENZ05215313
Leverancier:  Lenz Laborglas
Condensators, Davies
Condensators, Davies
250 mm, 24/29 NS, 24/29 NS
Price
155,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two GL 14 threaded plastic olives.
Documents
Specificaties
  • Afmetingen kern:
    24/29 NS
  • Mantellengte:
    250 mm
  • Huls:
    24/29 NS