Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Glass beads

Glass beads, solid, 5 mm
Catalog # LENZ05124005
Supplier:   Lenz Laborglas
Glass beads
Catalog # LENZ05124005
Supplier:  Lenz Laborglas
Glass beads
Glass beads

Glass beads, solid, 5 mm

Price
45,70 €
Log in for availability
1 * 1 kg
Product Details & Documents

Solid soda-lime glass.

Specifications
  • Ø:
    5 mm
  • Omschrijving:
    Glass beads, solid
  • Seq:
    04