Orders LinkContactUsLinkComponent
Adapters, 3 halzen
Adapters, 3 halzen
24/29 NS, 19/26 NS
Catalog # LENZ05015023
Supplier:   Lenz Laborglas
Adapters, 3 halzen
Catalog # LENZ05015023
Supplier:  Lenz Laborglas
Adapters, 3 halzen
Adapters, 3 halzen
24/29 NS, 19/26 NS
Price
71,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN® Borosilicate glass 3.3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).
Documents
Specifications
  • Afmetingen kern:
    24/29 NS
  • Huls:
    19/26 NS