Dropping funnels, conical shape, without pressure balance
2000 ml, 29/32 NS, 29/32 NS
Catalogus # 201-0917
Leverancier: Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Catalogus # 201-0917
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Borosilicate glass 3.3.
- Screw thread retaining device
- With inscription label, without graduation
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
BSE TSE Glass Columns
BSE/TSE-certificaten
Specificaties
- Afmetingen kern:29/32 NS
- Huls:29/32 NS
- Capaciteit:2000 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.