Orders LinkContactUsLinkComponent
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
2000 ml, 29/32 NS, 29/32 NS
Catalogus # 201-0917
Leverancier:   Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Catalogus # 201-0917
Leverancier:  Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
2000 ml, 29/32 NS, 29/32 NS
Price
233,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS
  • Capaciteit:
    2000 ml