Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Dropping funnels, conical shape, without pressure balance
1000 ml, 29/32 NS, 29/32 NS
Catalog # 532-0050
Supplier:   Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Catalog # 532-0050
Supplier:  Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
1000 ml, 29/32 NS, 29/32 NS
Price
145,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
Specifications
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS
  • Capaciteit:
    1000 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.