NS-stoppen
19/26, Flat
Catalog # 217-9094
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
- Excellent chemical resistance
- High thermal resistance, minimal heat expansion results in high resistance to temperature changes
- Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certifications: DIN 12252
Ordering information: Available with flat or pointed bottom.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- (NS):19/26
- Type:Flat
- Seq:14