Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
NS-stoppen
NS-stoppen
19/26, Flat
Catalog # 217-9094
Supplier:   Lenz Laborglas
NS-stoppen
Catalog # 217-9094
Supplier:  Lenz Laborglas
NS-stoppen
NS-stoppen
19/26, Flat
Price
48,80 €
Log in for availability
1 * 10 ST
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certifications: DIN 12252
Ordering information: Available with flat or pointed bottom.
Documents
Specifications
  • (NS):
    19/26
  • Type:
    Flat
  • Seq:
    14
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.