Orders LinkContactUsLinkComponent
Screw caps for screw thread tubes
Screw caps for screw thread tubes
GL 45
Catalogus # LENZ01320045
Leverancier:   Lenz Laborglas
Screw caps for screw thread tubes
Catalogus # LENZ01320045
Leverancier:  Lenz Laborglas
Screw caps for screw thread tubes
Screw caps for screw thread tubes
GL 45
Price
112,00 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents
PBT,red, with PTFE coated silicone sealing (VMQ).
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
Specificaties
  • Schroefdraad:
    GL 45
  • Seq:
    05