Adapters, PTFE, socket on cone
, volledige lengte, A, 14/35 NS, 10/30 NS, ,
Catalog # COWI021.01014
Supplier: COWIE
Jump to:
Product Details & Documents
Made of pure PTFE, white.
- Very good chemical resistance
- Very good temperature resistance
- Fully adjustable with replacement Viton® seal
- Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
Specifications
- Afmetingen kern:14/35 NS
- Huls:10/30 NS
- Type:volledige lengte, A
- Seq:01
- Cat. No.:COWI021.01014