Kwikdichloride ≥98%
Glass bottle for solids, 250 g
Leverancier: Thermo Fisher Scientific
Kwikdichloride ≥98%
Catalogus # A17472.30
Leverancier: Thermo Fisher Scientific
CAS-nummer: 7487-94-7
Ga naar:
Product Details & Documents
Purity: 98+% CAS No.: 7487-94-7 Molecular Formula: HgCl2 Formula Weight: 271.50 Refractive Index: 1.859 Odour: Odourless Boiling Point: 302° Merck Reference: 14,5876 UN No.: 1624 MDL No.: MFCD00011041
Synoniemen: Mercuric chloride, Mercury Chloride, Mercury dichloride
- CAS-nummer:7487-94-7
- Formule:HgCl₂
- MDL-nummer:MFCD00011041
- Molecular Weight:271.5 g/mol
- Merck index:14,5876
- Kokend Pt:302 °C
- Smelten Pt:280…282 °C
- Dichtheid:5,4
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:231-299-8
- UN:1624
- ADR:6.1,II
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:250 g



