Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Kwikdichloride ≥98%

Kwikdichloride ≥98%

Glass bottle for solids, 50 g
Catalogus # A17472.18
Giftig
Gezondheid
Corrosief
Milieu
Leverancier:   Thermo Fisher Scientific
Kwikdichloride ≥98%
Catalogus # A17472.18
CAS-nummer:  7487-94-7
Kwikdichloride ≥98%
Kwikdichloride ≥98%

Glass bottle for solids, 50 g

CAS-nummer:  7487-94-7
Price
60,80 €
Log in voor beschikbaarheid
1 * 50 g
Product Details & Documents

Purity: 98+% CAS No.: 7487-94-7 Molecular Formula: HgCl2 Formula Weight: 271.50 Refractive Index: 1.859 Odour: Odourless Boiling Point: 302° Merck Reference: 14,5876 UN No.: 1624 MDL No.: MFCD00011041

Synoniemen:  Mercuric chloride, Mercury Chloride, Mercury dichloride
  • CAS-nummer:7487-94-7
  • Formule:HgCl₂
  • MDL-nummer:MFCD00011041
  • Molecular Weight:271.5 g/mol
  • Merck index:14,5876
  • Kokend Pt:302 °C
  • Smelten Pt:280…282 °C
  • Dichtheid:5,4
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:231-299-8
  • UN:1624
  • ADR:6.1,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    50 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.