Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Triphosgene ≥98%

Triphosgene ≥98%

Glass bottle for solids, 100 g
Catalogus # A14932.22
Giftig
Corrosief
Leverancier:   Thermo Fisher Scientific
Triphosgene ≥98%
Catalogus # A14932.22
CAS-nummer:  32315-10-9
Triphosgene ≥98%
Triphosgene ≥98%

Glass bottle for solids, 100 g

CAS-nummer:  32315-10-9
Price
475,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 32315-10-9 Molecular Formula: C3Cl6O3 Formula Weight: 296.75 Storage Temperature: 2°C to 8°C Melting Point: 77-91° Boiling Point: 203-206° Beilstein Reference: 1787583 UN No.: 2928 MDL No.: MFCD00062848

Synoniemen:  Bis(trichloromethyl) carbonate
  • CAS-nummer:32315-10-9
  • Formule:C₃Cl₆O₃
  • MDL-nummer:MFCD00062848
  • Molecular Weight:296.75 g/mol
  • Kokend Pt:203…206 °C
  • Smelten Pt:77…91 °C
  • Dichtheid:1,600
  • EINECS:250-986-3
  • UN:2928
  • ADR:6.1,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g

Specificaties