Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Maleïnezuur ≥98%

Maleïnezuur ≥98%

Glass bottle for solids, 5.000 g
Catalogus # A14596.0I
Irriterend
Leverancier:   Thermo Fisher Scientific
Maleïnezuur ≥98%
Catalogus # A14596.0I
CAS-nummer:  110-16-7
Maleïnezuur ≥98%
Maleïnezuur ≥98%

Glass bottle for solids, 5.000 g

CAS-nummer:  110-16-7
Price
306,00 €
Log in voor beschikbaarheid
1 * 5.000 g
Product Details & Documents

Purity: 98+% CAS No.: 110-16-7 Molecular Formula: C4H4O4 Formula Weight: 116.07 Flash Point: 127°(260°F) Melting Point: 132-136° Beilstein Reference: 605762 Merck Reference: 14,5703 MDL No.: MFCD00063177

  • CAS-nummer:110-16-7
  • Formule:C₄H₄O₄
  • MDL-nummer:MFCD00063177
  • Molecular Weight:116.07 g/mol
  • Merck index:14,5703
  • Kokend Pt:256 °C (1013 hPa)
  • Smelten Pt:132…136 °C
  • Dichtheid:1,59
  • Flits Pt:127 °C (260 °F)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:203-742-5
  • UN:3261
  • ADR:8,III
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5.000 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.