Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

1,4-Dioxaan ≥99% stabilised

Glass bottle for solids, 2.500 g
Catalogus # A14188.0E
Ontvlambaar
Gezondheid
Irriterend
Leverancier:   Thermo Fisher Scientific
1,4-Dioxaan ≥99% stabilised
Catalogus # A14188.0E
CAS-nummer:  123-91-1
1,4-Dioxaan ≥99% stabilised
1,4-Dioxaan ≥99% stabilised

Glass bottle for solids, 2.500 g

CAS-nummer:  123-91-1
Price
137,00 €
Log in voor beschikbaarheid
1 * 2.500 g
Product Details & Documents

Stabilized with ca 5-10 ppm BHT

Purity: 99+% CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Refractive Index: 1.4220 Odour: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Synoniemen:  Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
  • CAS-nummer:123-91-1
  • Formule:C₄H₈O₂
  • MDL-nummer:MFCD00006571
  • Molecular Weight:88.11 g/mol
  • Merck index:14,3300
  • Kokend Pt:100…102 °C
  • Smelten Pt:11,8 °C
  • Dichtheid:1,034
  • Flits Pt:12 °C (54 °F)
  • Temperatuur bij opslag:Kamertemperatuur
  • UN:1165
  • ADR:3,II
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 5-10 ppm
  • EINECS:204-661-8
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    2.500 g

Specificaties