4'-Fluoroacetophenone ≥99%
Glass bottle for solids, 500 g
Leverancier: Thermo Fisher Scientific
4'-Fluoroacetophenone ≥99%
Catalogus # A13950.36
Leverancier: Thermo Fisher Scientific
CAS-nummer: 403-42-9
Ga naar:
Product Details & Documents
Purity: 99% CAS No.: 403-42-9 Molecular Formula: C8H7FO Formula Weight: 138.14 Flash Point: 71°(159°F) Refractive Index: 1.5110 Melting Point: 4° Boiling Point: 194-196° Beilstein Reference: 386013 MDL No.: MFCD00000354
Synoniemen: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- CAS-nummer:403-42-9
- Formule:C₈H₇FO
- MDL-nummer:MFCD00000354
- Molecular Weight:138.14 g/mol
- Kokend Pt:194…196 °C
- Smelten Pt:4 °C
- Dichtheid:1,141
- Flits Pt:71 °C (159 °F)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:206-960-9
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:500 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



