Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Xanthydrol ≥98%

Glass bottle for solids, 25 g
Catalogus # A13476.14
Leverancier:   Thermo Fisher Scientific
Xanthydrol ≥98%
Catalogus # A13476.14
CAS-nummer:  90-46-0
Xanthydrol ≥98%
Xanthydrol ≥98%

Glass bottle for solids, 25 g

CAS-nummer:  90-46-0
Price
244,00 €
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 90-46-0 Molecular Formula: C13H10O2 Formula Weight: 198.22 Storage Temperature: 2°C to 8°C Melting Point: 122-126° Beilstein Reference: 10395 MDL No.: MFCD00005057

Synoniemen:  9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
  • CAS-nummer:90-46-0
  • Formule:C₁₃H₁₀O₂
  • MDL-nummer:MFCD00005057
  • Molecular Weight:198.22 g/mol
  • Kokend Pt:338 °C (1013 hPa)
  • Smelten Pt:122…126 °C
  • EINECS:201-996-1
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.