Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Phthalaldehyde ≥98%

Phthalaldehyde ≥98%

Glass bottle for solids, 250 g
Catalogus # A13299.30
Giftig
Corrosief
Irriterend
Leverancier:   Thermo Fisher Scientific
Phthalaldehyde ≥98%
Catalogus # A13299.30
CAS-nummer:  643-79-8
Phthalaldehyde ≥98%
Phthalaldehyde ≥98%

Glass bottle for solids, 250 g

CAS-nummer:  643-79-8
Price
1 010,00 €
Log in voor beschikbaarheid
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 643-79-8 Molecular Formula: C8H6O2 Formula Weight: 134.14 Flash Point: 132°(269°F) Melting Point: 53-57° Beilstein Reference: 878317 UN No.: 2923 MDL No.: MFCD00003335

Synoniemen:  1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
  • CAS-nummer:643-79-8
  • Formule:C₈H₆O₂
  • MDL-nummer:MFCD00003335
  • Molecular Weight:134.13 g/mol
  • Kokend Pt:266 °C (1013 hPa)
  • Smelten Pt:53…57 °C
  • Dichtheid:1,13 g/cm³ (20 °C)
  • Flits Pt:132 °C (269 °F)
  • Temperatuur bij opslag:Koelkast
  • EINECS:211-402-2
  • UN:2923
  • ADR:8,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    250 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.