Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
2,3,5-Collidine ≥99%

2,3,5-Collidine ≥99%

Glass bottle for solids, 100 g
Catalogus # B23537.22
Giftig
Irriterend
Leverancier:   Thermo Fisher Scientific
2,3,5-Collidine ≥99%
Catalogus # B23537.22
CAS-nummer:  695-98-7
2,3,5-Collidine ≥99%
2,3,5-Collidine ≥99%

Glass bottle for solids, 100 g

CAS-nummer:  695-98-7
Price
199,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 695-98-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 74°(165°F) Refractive Index: 1.5075 Odour: Unpleasant Boiling Point: 182-184° Beilstein Reference: 108832 UN No.: 2810 MDL No.: MFCD00126840

Synoniemen:  2,3,5-Trimethylpyridine
  • CAS-nummer:695-98-7
  • Formule:C₈H₁₁N
  • MDL-nummer:MFCD00126840
  • Molecular Weight:121.18 g/mol
  • Kokend Pt:182…184 °C
  • Dichtheid:0,935
  • Flits Pt:74 °C (165 °F)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:211-786-1
  • UN:2810
  • ADR:6.1,III
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.