Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
2,3,5-Collidine ≥99%
Glass bottle for solids, 25 g
  B23537.14
2,3,5-Collidine ≥99%
  B23537.14
 :  695-98-7
2,3,5-Collidine ≥99%
2,3,5-Collidine ≥99%
Glass bottle for solids, 25 g
 :  695-98-7
72,30 €
1 * 25 g
Purity: 99% CAS No.: 695-98-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 74°(165°F) Refractive Index: 1.5075 Odour: Unpleasant Boiling Point: 182-184° Beilstein Reference: 108832 UN No.: 2810 MDL No.: MFCD00126840
 :  2,3,5-Trimethylpyridine
  •  :695-98-7
  •  :C₈H₁₁N
  •  :MFCD00126840
  •  :121.18 g/mol
  •  :182…184 °C
  •  :0,935
  •  :74 °C (165 °F)
  •  :Kamertemperatuur
  •  :211-786-1
  •  :2810
  •  :6.1,III
  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.