2,3,5-Collidine ≥99%
Glass bottle for solids, 25 g
Purity: 99% CAS No.: 695-98-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 74°(165°F) Refractive Index: 1.5075 Odour: Unpleasant Boiling Point: 182-184° Beilstein Reference: 108832 UN No.: 2810 MDL No.: MFCD00126840
: 2,3,5-Trimethylpyridine
- :695-98-7
- :C₈H₁₁N
- :MFCD00126840
- :121.18 g/mol
- :182…184 °C
- :0,935
- :74 °C (165 °F)
- :Kamertemperatuur
- :211-786-1
- :2810
- :6.1,III
- Verpakking:Glass bottle for solids
- VPE:25 g