Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
2,3,5-Collidine ≥99%

2,3,5-Collidine ≥99%

Glass bottle for solids, 5 g
Catalog # B23537.06
Toxic
Irritant
2,3,5-Collidine ≥99%
Catalog # B23537.06
CAS Number:  695-98-7
2,3,5-Collidine ≥99%
2,3,5-Collidine ≥99%

Glass bottle for solids, 5 g

CAS Number:  695-98-7
Price
34,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 695-98-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 74°(165°F) Refractive Index: 1.5075 Odour: Unpleasant Boiling Point: 182-184° Beilstein Reference: 108832 UN No.: 2810 MDL No.: MFCD00126840

Synonyms:  2,3,5-Trimethylpyridine
  • CAS Number:695-98-7
  • Formula:C₈H₁₁N
  • MDL Number:MFCD00126840
  • Molecular Weight:121.18 g/mol
  • Boiling Pt:182…184 °C
  • Density:0,935
  • Flash Pt:74 °C (165 °F)
  • Storage Temperature:Kamertemperatuur
  • EINECS:211-786-1
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.