Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Basic fuchsin

Basic fuchsin

Glass bottle for solids, 100 g
Catalogus # A12952.22
Gezondheid
Irriterend
Leverancier:   Thermo Fisher Scientific
Basic fuchsin
Catalogus # A12952.22
CAS-nummer:  632-99-5
Basic fuchsin
Basic fuchsin

Glass bottle for solids, 100 g

CAS-nummer:  632-99-5
Price
221,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents

CAS No.: 632-99-5 Molecular Formula: C20H20ClN3 Formula Weight: 337.86 Melting Point: 268-270° dec. Solubility: Soluble in water Beilstein Reference: 4166684 Merck Reference: 14,5652 MDL No.: MFCD00012569

Synoniemen:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS-nummer:632-99-5
  • Formule:C₂₀H₂₀ClN₃
  • MDL-nummer:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck index:14,5652
  • Smelten Pt:268…270 °C (ontbinding)
  • Dichtheid:1,22 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:211-189-6
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g

Specificaties