Ethyleendiamine 99%
Glass bottle, 500 mL
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Purity: 99% CAS No.: 107-15-3 Molecular Formula: C2H8N2 Formula Weight: 60.10 Flash Point: 38°(100°F) Physical Form: Liquid Density: 0.899 Refractive Index: 1.4565 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Odour: Ammonia-like Melting Point: 8-11° Boiling Point: 117-118° Solubility: Soluble in water, alcohol. Slightly soluble in ether Beilstein Reference: 605263 Merck Reference: 14,3795 UN No.: 1604 MDL No.: MFCD00008204
Strongly basic amine useful as a building block in chemical synthesis
Synoniemen: 1,2-Ethanediamine, 1,2-Diaminoethane
- CAS-nummer:107-15-3
- Formule:C₂H₈N₂
- MDL-nummer:MFCD00008204
- Molecular Weight:60.1 g/mol
- Merck index:14,3795
- Kokend Pt:117…118 °C
- Smelten Pt:8…11 °C
- Dichtheid:0,899
- Flits Pt:38 °C (100 °F)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:203-468-6
- UN:1604
- ADR:8,II
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle
- VPE:500 mL
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



