Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Ethyleendiamine 99%
Glass bottle, 100 mL
  A12132.AE
Ethyleendiamine 99%
  A12132.AE
 :  107-15-3
Ethyleendiamine 99%
Ethyleendiamine 99%
Glass bottle, 100 mL
 :  107-15-3
31,00 €
1 * 100 mL
Purity: 99% CAS No.: 107-15-3 Molecular Formula: C2H8N2 Formula Weight: 60.10 Flash Point: 38°(100°F) Physical Form: Liquid Density: 0.899 Refractive Index: 1.4565 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Odour: Ammonia-like Melting Point: 8-11° Boiling Point: 117-118° Solubility: Soluble in water, alcohol. Slightly soluble in ether Beilstein Reference: 605263 Merck Reference: 14,3795 UN No.: 1604 MDL No.: MFCD00008204
Strongly basic amine useful as a building block in chemical synthesis
 :  1,2-Ethanediamine, 1,2-Diaminoethane
  •  :107-15-3
  •  :C₂H₈N₂
  •  :MFCD00008204
  •  :60.1 g/mol
  •  :14,3795
  •  :117…118 °C
  •  :8…11 °C
  •  :0,899
  •  :38 °C (100 °F)
  •  :Kamertemperatuur
  •  :203-468-6
  •  :1604
  •  :8,II
  • Verpakking:
    Glass bottle
  • VPE:
    100 mL