Ethyleendiamine 99%
Glass bottle, 100 mL
Purity: 99% CAS No.: 107-15-3 Molecular Formula: C2H8N2 Formula Weight: 60.10 Flash Point: 38°(100°F) Physical Form: Liquid Density: 0.899 Refractive Index: 1.4565 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Odour: Ammonia-like Melting Point: 8-11° Boiling Point: 117-118° Solubility: Soluble in water, alcohol. Slightly soluble in ether Beilstein Reference: 605263 Merck Reference: 14,3795 UN No.: 1604 MDL No.: MFCD00008204
Strongly basic amine useful as a building block in chemical synthesis
: 1,2-Ethanediamine, 1,2-Diaminoethane
- :107-15-3
- :C₂H₈N₂
- :MFCD00008204
- :60.1 g/mol
- :14,3795
- :117…118 °C
- :8…11 °C
- :0,899
- :38 °C (100 °F)
- :Kamertemperatuur
- :203-468-6
- :1604
- :8,II
- Verpakking:Glass bottle
- VPE:100 mL