Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Ethyleendiamine 99%

Plastic bottle, 10.000 mL
Catalogus # A12132.0D
Gezondheid
Corrosief
Ontvlambaar
Irriterend
Leverancier:   Thermo Fisher Scientific
Ethyleendiamine 99%
Catalogus # A12132.0D
CAS-nummer:  107-15-3
Ethyleendiamine 99%
Ethyleendiamine 99%

Plastic bottle, 10.000 mL

CAS-nummer:  107-15-3
Price
342,00 €
Log in voor beschikbaarheid
1 * 10.000 mL
Product Details & Documents

Purity: 99% CAS No.: 107-15-3 Molecular Formula: C2H8N2 Formula Weight: 60.10 Flash Point: 38°(100°F) Physical Form: Liquid Density: 0.899 Refractive Index: 1.4565 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Odour: Ammonia-like Melting Point: 8-11° Boiling Point: 117-118° Solubility: Soluble in water, alcohol. Slightly soluble in ether Beilstein Reference: 605263 Merck Reference: 14,3795 UN No.: 1604 MDL No.: MFCD00008204

Strongly basic amine useful as a building block in chemical synthesis

Synoniemen:  1,2-Ethanediamine, 1,2-Diaminoethane
  • CAS-nummer:107-15-3
  • Formule:C₂H₈N₂
  • MDL-nummer:MFCD00008204
  • Molecular Weight:60.1 g/mol
  • Merck index:14,3795
  • Kokend Pt:117…118 °C
  • Smelten Pt:8…11 °C
  • Dichtheid:0,899
  • Flits Pt:38 °C (100 °F)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:203-468-6
  • UN:1604
  • ADR:8,II
Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    10.000 mL

Specificaties