Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

trans-2-Hydroxycinnamic acid ≥98%

Glass bottle for solids, 100 g
Catalog # A11740.22
Irritant
trans-2-Hydroxycinnamic acid ≥98%
Catalog # A11740.22
CAS Number:  614-60-8
trans-2-Hydroxycinnamic acid ≥98%
trans-2-Hydroxycinnamic acid ≥98%

Glass bottle for solids, 100 g

CAS Number:  614-60-8
Price
269,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 614-60-8 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: ca 210° dec. Beilstein Reference: 1100900

Synonyms:  (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid
  • CAS Number:614-60-8
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00004379
  • Molecular Weight:164.16 g/mol
  • Boiling Pt:330 °C (1013 hPa)
  • Melting Pt:∼ 210 °C (ontbinding)
  • Storage Temperature:Kamertemperatuur
  • EINECS:210-386-4
Specifications

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g

Specifications