Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
trans-2-Hydroxycinnamic acid ≥98%
Glass bottle for solids, 25 g
  A11740.14
trans-2-Hydroxycinnamic acid ≥98%
  A11740.14
 :  614-60-8
trans-2-Hydroxycinnamic acid ≥98%
trans-2-Hydroxycinnamic acid ≥98%
Glass bottle for solids, 25 g
 :  614-60-8
87,90 €
1 * 25 g
Purity: 98+% CAS No.: 614-60-8 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: ca 210° dec. Beilstein Reference: 1100900
 :  (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid
  •  :614-60-8
  •  :C₉H₈O₃
  •  :MFCD00004379
  •  :164.16 g/mol
  •  :330 °C (1013 hPa)
  •  :∼ 210 °C (ontbinding)
  •  :Kamertemperatuur
  •  :210-386-4
  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g