trans-2-Hydroxycinnamic acid ≥98%
Glass bottle for solids, 25 g
Purity: 98+% CAS No.: 614-60-8 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: ca 210° dec. Beilstein Reference: 1100900
: (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid
- :614-60-8
- :C₉H₈O₃
- :MFCD00004379
- :164.16 g/mol
- :330 °C (1013 hPa)
- :∼ 210 °C (ontbinding)
- :Kamertemperatuur
- :210-386-4
- Verpakking:Glass bottle for solids
- VPE:25 g