Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Malic acid ≥98%

D-(+)-Malic acid ≥98%

Glass bottle for solids, 5 g
Catalogus # A11688.06
Corrosief
Irriterend
Leverancier:   Thermo Fisher Scientific
D-(+)-Malic acid ≥98%
Catalogus # A11688.06
CAS-nummer:  636-61-3
D-(+)-Malic acid ≥98%
D-(+)-Malic acid ≥98%

Glass bottle for solids, 5 g

CAS-nummer:  636-61-3
Price
80,20 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 636-61-3 Molecular Formula: C4H6O5 Formula Weight: 134.09 Melting Point: 100-104° Beilstein Reference: 1723540 Merck Reference: 14,5707 Optical Rotation: +28° (c=5.5 in pyridine) MDL No.: MFCD00004245

Synoniemen:  (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
  • CAS-nummer:636-61-3
  • Formule:C₄H₆O₅
  • MDL-nummer:MFCD00004245
  • Molecular Weight:134.09 g/mol
  • Merck index:14,5707
  • Kokend Pt:322 °C (1013 hPa)
  • Smelten Pt:100…104 °C
  • Dichtheid:1,60
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:211-262-2
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.